6-Fluoro-8-methylquinoline ≥97%

Supplier: AOBChem USA

Warning

121461-5G 121461-25G 121461-1G 121461-500MG
77633-954EA 304 USD
77633-954 77653-346 77640-466 77644-450
6-Fluoro-8-methylquinoline ≥97%
6-Fluoro-8-methylquinoline

Precaución: For Research Use Only

Formula: C₁₀H₈FN
MW: 161.176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026038
CAS Number: 1150271-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR