3-Chloro-4-cyanobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

121233-5G 121233-1G 121233-25G
77633-952EA 208 USD
77633-952 77636-446 77657-288
3-Chloro-4-cyanobenzoic acid ≥97%
3-Chloro-4-cyanobenzoic acid

Precaución: For Research Use Only

Formula: C₈H₄ClNO₂
MW: 181.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18392433
CAS Number: 1261685-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR