1-(3,4-Dimethylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

120943-5G 120943-1G 120943-25G 120943-500MG
77633-950EA 392 USD
77633-950 77636-444 77653-342 77648-452
1-(3,4-Dimethylphenyl)ethanol ≥95%
1-(3,4-Dimethylphenyl)ethanol

Precaución: For Research Use Only

Formula: 150.2175 g/mol
MW: C₁₀H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD08442206
CAS Number: 33967-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR