5-Bromo-2-ethyl-2H-indazole ≥97%

Supplier: AOBChem USA

Warning

120901-5G 120901-25G 120901-1G 120901-500MG
77633-948EA 458 USD
77633-948 77657-286 77644-448 77648-450
5-Bromo-2-ethyl-2H-indazole ≥97%
5-Bromo-2-ethyl-2H-indazole

Precaución: For Research Use Only

Formula: C₉H₉BrN₂
MW: 225.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18434483
CAS Number: 590417-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR