5-Bromo-1-ethyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

120895-5G 120895-1G 120895-25G 120895-500MG
77631-524EA 671 USD
77631-524 77636-442 77653-340 77644-446
5-Bromo-1-ethyl-1H-indazole ≥97%
5-Bromo-1-ethyl-1H-indazole

Precaución: For Research Use Only

Formula: C₉H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD17010084
CAS Number: 590417-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR