6-Fluoro-2,3-dihydrobenzo[b][1,4]dioxine ≥95%

Supplier: AOBChem USA

Warning

120698-5G 120698-25G 120698-1G 120698-500MG
77631-522EA 1228 USD
77631-522 77657-284 77640-464 77644-444
6-Fluoro-2,3-dihydrobenzo[b][1,4]dioxine ≥95%
6-Fluoro-2,3-dihydrobenzo[b][1,4]dioxine

Precaución: For Research Use Only

Formula: 154.138 g/mol
MW: C₈H₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD27962182
CAS Number: 60458-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR