1-(3-Methoxy-4-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

120580-5G 120580-1G 120580-25G 120580-500MG
77631-128EA 1678.9 USD
77631-128 77636-440 77657-282 77648-448
1-(3-Methoxy-4-methylphenyl)cyclobutanol ≥95%
1-(3-Methoxy-4-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 192.254 g/mol
MW: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD34824123
CAS Number: 2969334-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR