2-(3-Methoxy-4-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

120576-5G 120576-25G 120576-1G 120576-500MG
77631-520EA 1171 USD
77631-520 77653-338 77644-442 77648-446
2-(3-Methoxy-4-methylphenyl)propan-2-ol ≥95%
2-(3-Methoxy-4-methylphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 180.244 g/mol
MW: C₁₁H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD28357468
CAS Number: 40180-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR