2-Chloro-4-iodo-1-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-4-iodo-1-(trifluoromethyl)benzene

120038-5G 120038-1G 120038-500MG 120038-25G
77631-518EA 884 USD
77631-518 77636-436 77652-308 77653-334
2-Chloro-4-iodo-1-(trifluoromethyl)benzene ≥97%
2-Chloro-4-iodo-1-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: C₇H₃ClF₃I
MW: 306.451 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12827841
CAS Number: 1206599-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR