1-Bromo-3-cyclopentyl-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

119974-5G 119974-1G 119974-500MG
77653-332EA 2314 USD
77653-332 77640-462 77648-442
1-Bromo-3-cyclopentyl-2-fluorobenzene ≥97%
1-Bromo-3-cyclopentyl-2-fluorobenzene

Precaución: For Research Use Only

Formula: 243.115 g/mol
MW: C₁₁H₁₂BrF
Storage Temperature: Ambient
MDL Number: MFCD26678744
CAS Number: 1513525-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR