1-(2,6-Dichloro-4-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

119630-5G 119630-1G 119630-25G 119630-500MG
77631-122EA 1557.36 USD
77631-122 77636-432 77653-328 77644-438
1-(2,6-Dichloro-4-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone ≥97%
1-(2,6-Dichloro-4-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 311.008 g/mol
MW: C₉H₂Cl₂F₆O
Storage Temperature: Ambient
MDL Number: MFCD34823825
CAS Number: 2921835-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR