1-(2-Chloro-3-methylphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

119491-5G 119491-1G 119491-500MG
77631-120EA 1484.23 USD
77631-120 77636-430 77648-438
1-(2-Chloro-3-methylphenyl)ethanol ≥97%
1-(2-Chloro-3-methylphenyl)ethanol

Precaución: For Research Use Only

Formula: C₉H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD30081219
CAS Number: 1593008-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR