5-Bromo-2-methoxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

119429-5G 119429-500MG 119429-25G 119429-1G
77631-510EA 1381 USD
77631-510 77652-306 77653-326 77640-456
5-Bromo-2-methoxy-3-methylbenzoic acid ≥97%
5-Bromo-2-methoxy-3-methylbenzoic acid

Precaución: For Research Use Only

Formula: 245.07 g/mol
MW: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD05797690
CAS Number: 159121-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR