4-Bromo-1-cyclohexyl-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

119037-5G 119037-25G 119037-1G 119037-500MG
77631-116EA 1484.23 USD
77631-116 77635-416 77636-428 77648-436
4-Bromo-1-cyclohexyl-2-methylbenzene ≥97%
4-Bromo-1-cyclohexyl-2-methylbenzene

Precaución: For Research Use Only

Formula: C₁₃H₁₇Br
Storage Temperature: Ambient
MDL Number: MFCD35165886
CAS Number: 343334-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR