3,4-Dibromo-5-iodobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

118958-5G 118958-1G 118958-500MG 118958-25G
77631-114EA 1484.23 USD
77631-114 77636-426 77652-304 77657-268
3,4-Dibromo-5-iodobenzoic acid ≥97%
3,4-Dibromo-5-iodobenzoic acid

Precaución: For Research Use Only

Formula: C₇H₃Br₂IO₂
Storage Temperature: Ambient
MDL Number: MFCD34784264
CAS Number: 2166915-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR