(3-Chloro-2,6-difluoro-5-iodophenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

118948-5G 118948-1G 118948-25G 118948-500MG
77631-508EA 1441 USD
77631-508 77636-424 77657-266 77644-432
(3-Chloro-2,6-difluoro-5-iodophenyl)(methyl)sulfane ≥95%
(3-Chloro-2,6-difluoro-5-iodophenyl)(methyl)sulfane

Precaución: For Research Use Only

Formula: C₇H₄ClF₂IS
Storage Temperature: Ambient
MDL Number: MFCD34784257
CAS Number: 2918924-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR