1-Fluoro-2-iodo-3-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

118928-5G 118928-1G 118928-500MG
77657-262EA 2417 USD
77657-262 77640-450 77648-432
1-Fluoro-2-iodo-3-propoxybenzene ≥95%
1-Fluoro-2-iodo-3-propoxybenzene

Precaución: For Research Use Only

Formula: C₉H₁₀FIO
MW: 280.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34784245
CAS Number: 1872784-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR