3-Bromo-6-chloro-2,4-difluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

118859-5G 118859-500MG 118859-25G 118859-1G
77631-502EA 1184 USD
77631-502 77652-302 77653-322 77640-448
3-Bromo-6-chloro-2,4-difluorobenzaldehyde ≥95%
3-Bromo-6-chloro-2,4-difluorobenzaldehyde

Precaución: For Research Use Only

Formula: 255.444 g/mol
MW: C₇H₂BrClF₂O
Storage Temperature: Ambient
CAS Number: 1160573-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR