(3,4-Difluoro-2-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118545-5G 118545-1G 118545-25G 118545-500MG
77631-498EA 1441 USD
77631-498 77636-416 77657-258 77644-426
(3,4-Difluoro-2-methylphenyl)(phenyl)methanol ≥95%
(3,4-Difluoro-2-methylphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 234.241 g/mol
MW: C₁₄H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD34784149
CAS Number: 2918858-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR