(2-Methyl-4-(trifluoromethyl)phenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118538-5G 118538-1G 118538-25G 118538-500MG
77631-496EA 1441 USD
77631-496 77636-414 77653-320 77648-428
(2-Methyl-4-(trifluoromethyl)phenyl)(phenyl)methanol ≥95%
(2-Methyl-4-(trifluoromethyl)phenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: C₁₅H₁₃F₃O
Storage Temperature: Ambient
MDL Number: MFCD34784142
CAS Number: 1869063-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR