(2,4-Bis(trifluoromethyl)phenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118536-5G 118536-25G 118536-1G 118536-500MG
77631-494EA 1441 USD
77631-494 77657-256 77640-446 77648-426
(2,4-Bis(trifluoromethyl)phenyl)(phenyl)methanol ≥95%
(2,4-Bis(trifluoromethyl)phenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 320.23 g/mol
MW: C₁₅H₁₀F₆O
Storage Temperature: Ambient
MDL Number: MFCD34784141
CAS Number: 2918838-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR