(3,5-Bis(trifluoromethyl)phenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118535-5G 118535-25G 118535-1G 118535-500MG
77631-492EA 1078 USD
77631-492 77653-318 77640-444 77644-424
(3,5-Bis(trifluoromethyl)phenyl)(phenyl)methanol ≥95%
(3,5-Bis(trifluoromethyl)phenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: C₁₅H₁₀F₆O
Storage Temperature: Ambient
MDL Number: MFCD11100929
CAS Number: 81390-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR