(4-Chloro-2-methoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118527-5G 118527-500MG 118527-25G 118527-1G
77631-490EA 1441 USD
77631-490 77652-300 77653-316 77640-442
(4-Chloro-2-methoxyphenyl)(phenyl)methanol ≥95%
(4-Chloro-2-methoxyphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 248.705 g/mol
MW: C₁₄H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD16507402
CAS Number: 1280642-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR