(2-Chloro-4-methoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118519-5G 118519-1G 118519-25G 118519-500MG
77631-488EA 1441 USD
77631-488 77636-412 77653-314 77648-424
(2-Chloro-4-methoxyphenyl)(phenyl)methanol ≥95%
(2-Chloro-4-methoxyphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 248.705 g/mol
MW: C₁₄H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD16359519
CAS Number: 1307244-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR