(3-Chloro-4-(trifluoromethyl)phenyl)(phenyl)methanol ≥95%
Supplier: AOBChem USA
|
Warning
|
118518-5G
118518-25G
118518-1G
118518-500MG
77631-486EA
1441
USD
77631-486
77657-254
77640-440
77644-422
(3-Chloro-4-(trifluoromethyl)phenyl)(phenyl)methanol ≥95%
(3-Chloro-4-(trifluoromethyl)phenyl)(phenyl)methanol
Precaución: For Research Use Only
Formula:
C₁₄H₁₀ClF₃O Storage Temperature: Ambient |
MDL Number:
MFCD34784128 CAS Number: 2918924-52-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...