(3-Chloro-4-methoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118510-5G 118510-25G 118510-1G 118510-500MG
77631-484EA 1441 USD
77631-484 77653-310 77640-436 77644-420
(3-Chloro-4-methoxyphenyl)(phenyl)methanol ≥95%
(3-Chloro-4-methoxyphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: C₁₄H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD16356770
CAS Number: 1281727-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR