(3-Fluoro-2-methoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118509-5G 118509-25G 118509-1G 118509-500MG
77631-482EA 1441 USD
77631-482 77657-252 77640-434 77644-418
(3-Fluoro-2-methoxyphenyl)(phenyl)methanol ≥95%
(3-Fluoro-2-methoxyphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 232.25 g/mol
MW: C₁₄H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34784123
CAS Number: 2918844-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR