(3-Chloro-4-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118503-5G 118503-1G 118503-25G 118503-500MG
77631-478EA 1441 USD
77631-478 77636-408 77653-308 77648-418
(3-Chloro-4-methylphenyl)(phenyl)methanol ≥95%
(3-Chloro-4-methylphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 232.705 g/mol
MW: C₁₄H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD18287275
CAS Number: 1343599-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR