(3-Chloro-2-methoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118500-5G 118500-1G 118500-25G 118500-500MG
77631-474EA 1441 USD
77631-474 77636-406 77653-304 77648-414
(3-Chloro-2-methoxyphenyl)(phenyl)methanol ≥95%
(3-Chloro-2-methoxyphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 248.705 g/mol
MW: C₁₄H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34784121
CAS Number: 1419567-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR