(2,3-Dimethylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118481-5G 118481-1G 118481-25G 118481-500MG
77631-468EA 1441 USD
77631-468 77636-402 77657-248 77648-410
(2,3-Dimethylphenyl)(phenyl)methanol ≥95%
(2,3-Dimethylphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 212.287 g/mol
MW: C₁₅H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD12786890
CAS Number: 21945-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR