(3,4-Dimethylphenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

118478-5G 118478-25G 118478-1G 118478-500MG
77631-466EA 1284 USD
77631-466 77653-298 77640-428 77644-414
(3,4-Dimethylphenyl)(phenyl)methanol ≥97%
(3,4-Dimethylphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: C₁₅H₁₆O
MW: 212.287 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08443131
CAS Number: 14189-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR