(4-Phenoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118466-5G 118466-500MG 118466-1G
77631-108EA 1484.23 USD
77631-108 77652-298 77640-426
(4-Phenoxyphenyl)(phenyl)methanol ≥95%
(4-Phenoxyphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 276.329 g/mol
MW: C₁₉H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD11096257
CAS Number: 34979-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR