(2-Ethoxyphenyl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

118428-5G 118428-1G 118428-25G 118428-500MG
77631-464EA 1441 USD
77631-464 77636-400 77657-246 77648-408
(2-Ethoxyphenyl)(phenyl)methanol ≥97%
(2-Ethoxyphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: C₁₅H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD16314670
CAS Number: 94001-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR