1-(3-Chloro-2-methoxypyridin-4-yl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

118317-5G 118317-1G 118317-25G 118317-500MG
77631-106EA 1557.36 USD
77631-106 77636-396 77653-296 77644-410
1-(3-Chloro-2-methoxypyridin-4-yl)-2-methylpropan-1-ol ≥95%
1-(3-Chloro-2-methoxypyridin-4-yl)-2-methylpropan-1-ol

Precaución: For Research Use Only

Formula: 215.677 g/mol
MW: C₁₀H₁₄ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD34784061
CAS Number: 2169342-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR