(5-Chloro-2,3-difluoropyridin-4-yl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

118255-5G 118255-1G 118255-25G 118255-500MG
77631-102EA 1484.23 USD
77631-102 77636-394 77657-240 77648-406
(5-Chloro-2,3-difluoropyridin-4-yl)(phenyl)methanol ≥97%
(5-Chloro-2,3-difluoropyridin-4-yl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 255.648 g/mol
MW: C₁₂H₈ClF₂NO
Storage Temperature: Ambient
MDL Number: MFCD34784004
CAS Number: 2918837-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR