(6-Chloro-2-fluoro-3-iodophenyl)(phenyl)methanol ≥95%
Supplier: AOBChem USA
|
Warning
|
118246-5G
118246-500MG
118246-25G
118246-1G
77631-098EA
1484.23
USD
77631-098
77652-296
77653-294
77640-420
(6-Chloro-2-fluoro-3-iodophenyl)(phenyl)methanol ≥95%
(6-Chloro-2-fluoro-3-iodophenyl)(phenyl)methanol
Precaución: For Research Use Only
Formula:
362.566 g/mol MW: C₁₃H₉ClFIO Storage Temperature: Ambient |
MDL Number:
MFCD34783997 CAS Number: 2918912-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...