(5-Chloro-2-fluoro-3-methoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118210-5G 118210-25G 118210-1G 118210-500MG
77631-096EA 1589.29 USD
77631-096 77653-292 77640-418 77648-402
(5-Chloro-2-fluoro-3-methoxyphenyl)(phenyl)methanol ≥95%
(5-Chloro-2-fluoro-3-methoxyphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 266.695 g/mol
MW: C₁₄H₁₂ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD34783962
CAS Number: 2918872-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR