(2-Fluoro-3-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118204-5G 118204-25G 118204-1G 118204-500MG
77631-462EA 1381 USD
77631-462 77657-238 77640-416 77644-406
(2-Fluoro-3-methylphenyl)(phenyl)methanol ≥95%
(2-Fluoro-3-methylphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 216.251 g/mol
MW: C₁₄H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD24115536
CAS Number: 1512584-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR