(5-Bromo-2-fluoro-3-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118200-5G 118200-1G 118200-25G 118200-500MG
77631-094EA 1484.23 USD
77631-094 77636-392 77653-290 77644-404
(5-Bromo-2-fluoro-3-methylphenyl)(phenyl)methanol ≥95%
(5-Bromo-2-fluoro-3-methylphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 295.147 g/mol
MW: C₁₄H₁₂BrFO
Storage Temperature: Ambient
MDL Number: MFCD34783955
CAS Number: 2918863-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR