2,6-Difluoro-3-(hydroxy(phenyl)methyl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

118199-5G 118199-25G 118199-1G 118199-500MG
77631-092EA 1707.74 USD
77631-092 77657-236 77640-414 77644-402
2,6-Difluoro-3-(hydroxy(phenyl)methyl)benzonitrile ≥95%
2,6-Difluoro-3-(hydroxy(phenyl)methyl)benzonitrile

Precaución: For Research Use Only

Formula: 245.224 g/mol
MW: C₁₄H₉F₂NO
Storage Temperature: Ambient
MDL Number: MFCD34783954
CAS Number: 2918931-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR