(2,3-Dichloropyridin-4-yl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118196-5G 118196-25G 118196-1G 118196-500MG
77631-460EA 1441 USD
77631-460 77653-288 77640-412 77644-400
(2,3-Dichloropyridin-4-yl)(phenyl)methanol ≥95%
(2,3-Dichloropyridin-4-yl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 254.112 g/mol
MW: C₁₂H₉Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD34783952
CAS Number: 2918892-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR