(2-Chloro-3-fluoropyridin-4-yl)(phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

118174-5G 118174-25G 118174-1G 118174-500MG
77631-458EA 1441 USD
77631-458 77653-286 77640-410 77644-398
(2-Chloro-3-fluoropyridin-4-yl)(phenyl)methanol ≥97%
(2-Chloro-3-fluoropyridin-4-yl)(phenyl)methanol

Precaución: For Research Use Only

Formula: C₁₂H₉ClFNO
MW: 237.657 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34783930
CAS Number: 2918931-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR