(6-Bromo-2-chloro-3-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

118171-5G 118171-25G 118171-1G 118171-500MG
77631-090EA 1718.04 USD
77631-090 77657-234 77640-408 77648-400
(6-Bromo-2-chloro-3-methylphenyl)(phenyl)methanol ≥95%
(6-Bromo-2-chloro-3-methylphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 311.601 g/mol
MW: C₁₄H₁₂BrClO
Storage Temperature: Ambient
MDL Number: MFCD34783927
CAS Number: 2918924-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR