2-Chloro-2,2-difluoro-1-(2-fluoro-6-(trifluoromethyl)phenyl)ethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
117737-5G
117737-25G
117737-1G
117737-500MG
77631-088EA
1484.23
USD
77631-088
77657-232
77640-406
77648-398
2-Chloro-2,2-difluoro-1-(2-fluoro-6-(trifluoromethyl)phenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(2-fluoro-6-(trifluoromethyl)phenyl)ethanone
Precaución: For Research Use Only
Formula:
C₉H₃ClF₆O Storage Temperature: Ambient |
MDL Number:
MFCD34823510 CAS Number: 2969368-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...