Ethyl 2-(3-bromo-2-fluoro-6-iodophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

117728-5G 117728-25G 117728-1G 117728-500MG
77631-086EA 1616.07 USD
77631-086 77635-410 77640-404 77648-396
Ethyl 2-(3-bromo-2-fluoro-6-iodophenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-bromo-2-fluoro-6-iodophenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 400.9676 g/mol
MW: C₁₀H₇BrFIO₃
Storage Temperature: Ambient
MDL Number: MFCD34823508
CAS Number: 2969218-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR