4-Bromo-2-fluorothiobenzamide ≥97%

Supplier: AOBChem USA

Warning

117641-5G 117641-1G 117641-500MG
77631-082EA 1484.23 USD
77631-082 77640-400 77644-396
4-Bromo-2-fluorothiobenzamide ≥97%
4-Bromo-2-fluorothiobenzamide

Precaución: For Research Use Only

Formula: C₇H₅BrFNS
Storage Temperature: Ambient
MDL Number: MFCD08460505
CAS Number: 883230-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR