Methyl 4-cyano-5-fluoro-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

117512-5G 117512-1G 117512-25G 117512-500MG
77631-456EA 1398 USD
77631-456 77636-390 77653-282 77644-394
Methyl 4-cyano-5-fluoro-2-methylbenzoate ≥97%
Methyl 4-cyano-5-fluoro-2-methylbenzoate

Precaución: For Research Use Only

Formula: C₁₀H₈FNO₂
MW: 193.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23707556
CAS Number: 1427377-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR