4-Cyano-5-fluoro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

117511-5G 117511-25G 117511-1G 117511-500MG
77631-080EA 1484.23 USD
77631-080 77635-408 77640-398 77644-392
4-Cyano-5-fluoro-2-methylbenzoic acid ≥97%
4-Cyano-5-fluoro-2-methylbenzoic acid

Precaución: For Research Use Only

Formula: 179.148 g/mol
MW: C₉H₆FNO₂
Storage Temperature: Ambient
MDL Number: MFCD28769461
CAS Number: 1807296-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR