1-(2-Bromo-6-fluoro-4-(trifluoromethyl)phenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

117470-5G 117470-25G 117470-1G 117470-500MG
77631-076EA 1489.38 USD
77631-076 77653-278 77640-396 77648-392
1-(2-Bromo-6-fluoro-4-(trifluoromethyl)phenyl)-2-methylpropan-1-ol ≥95%
1-(2-Bromo-6-fluoro-4-(trifluoromethyl)phenyl)-2-methylpropan-1-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₁BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD34783503
CAS Number: 2918939-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR