1-(3-Bromo-2-fluorophenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

117461-5G 117461-1G 117461-25G 117461-500MG
77631-074EA 1484.23 USD
77631-074 77636-388 77653-276 77644-388
1-(3-Bromo-2-fluorophenyl)-2-methylpropan-1-ol ≥95%
1-(3-Bromo-2-fluorophenyl)-2-methylpropan-1-ol

Precaución: For Research Use Only

Formula: 247.104 g/mol
MW: C₁₀H₁₂BrFO
Storage Temperature: Ambient
MDL Number: MFCD30077387
CAS Number: 1528836-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR